ART™ Reload System
Actual product may vary
ART™ Reload System
Reduce waste and save benchtop and storage space with this advanced nonfiltered reload system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)For Use With (Equipment)Tip StyleFit for Purpose
3502-RT10 μLUniversal FitStandardCertified Quality
3101-05-RT1000 μLUniversal FitLow RetentionSterile
3101-RT1000 μLUniversal FitStandardSterile
3102-RT1000 μLUniversal FitStandardCertified Quality
3102-05-RT1000 μLUniversal FitLow RetentionCertified Quality
3501-05-RT10 μLUniversal FitLow RetentionSterile
3501-RT10 μLUniversal FitStandardSterile
3502-05-RT10 μLUniversal FitLow RetentionCertified Quality
3521-05-RT20 μLEppendorfLow RetentionSterile
3521-RT20 μLEppendorfStandardSterile
3522-05-RT20 μLEppendorfLow RetentionCertified Quality
3522-RT20 μLEppendorfStandardCertified Quality
3551-05-RT200 μLUniversal FitLow RetentionSterile
3551-RT200 μLUniversal FitStandardSterile
3552-05-RT200 μLUniversal FitLow RetentionCertified Quality
3552-RT200 μLUniversal FitStandardCertified Quality
3771-05-RT300 μLUniversal FitLow RetentionSterile
3771-RT300 μLUniversal FitStandardSterile
3772-05-RT300 μLUniversal FitLow RetentionCertified Quality
3772-RT300 μLUniversal FitStandardCertified Quality
Catalog number 3502-RT
HSN 84799090
Price (INR)
8,632.00
PK
Volume (Metric):
10 μL
For Use With (Equipment):
Universal Fit
Tip Style:
Standard
Fit for Purpose:
Certified Quality
Price (INR)
8,632.00
PK
Reduce waste and save benchtop and storage space with the advanced Thermo Scientific™ ART™ Nonfiltered Reload System. This system provides an environmentally responsible option without compromising sterility.
Features:
  • Stackable barrier tip packaging allows for tips to pass through offset holes in lower insert
  • Blister sleeve guide rails align tip inserts for smooth transfer into the rack
  • DNase, RNase, DNA, ATP, and Endotoxin Free
  • Outer sleeve protects tips from exterior contamination while refilling inserts
  • Tip packaging option reduces plastic waste
  • Rigid insert design provides reliable tip loading
  • Lot number and description are printed on every insert
Note:
Empty rack required for -RT parts
Specifications
Volume (Metric)10 μL
FilteredNon-filtered
SterilityNon-sterile
ColorNatural
ModelART Ultra Micro Non-Filtered
Length (English)1.2 in.
Length (Metric)3.13 cm
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
FormatRack
Tip StyleStandard
Quantity4800/Cs.
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeCertified Quality
Unit SizePK