2-Hydroxy-5-bromopyridine, 99%
2-Hydroxy-5-bromopyridine, 99%
2-Hydroxy-5-bromopyridine, 99%
Acros Organics

2-Hydroxy-5-bromopyridine, 99%

CAS: 13466-38-1 | C5H4BrNO | 174 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 357640250
also known as 35764
HSN 29333919
Price (INR)
36,478.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
36,478.00
Each
Chemical Identifiers
CAS13466-38-1
IUPAC Name5-bromo-1,2-dihydropyridin-2-one
Molecular FormulaC5H4BrNO
InChI KeyNDMZZQRNZFWMEZ-UHFFFAOYSA-N
SMILESBrC1=CNC(=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=98.5 %
Melting point179°C to 182°C
Appearance (Color)Off-white to yellow-brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only