Product Image
Thermo Scientific™
Accessories for Shaking Water Baths
Enhance flexibility using these accessories for shaking water baths.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
3582-37Cooling Coil
3582-33Platform Insert
3582-35Gassing Hood
3582-39Gable Cover
Catalog number 3582-37
Price (INR)
-
Type:
Cooling Coil
Enhance flexibility using Thermo Scientific™ Accessories for Shaking Water Baths.
  • Platform insert and one or more flask clamps are required to hold flasks in place
  • Gable cover extends temperature range to 98°C
  • Warranty varies by country, please contact us for details
Specifications
For Use With (Equipment)Analog Reciprocal Shaking Water Baths Models 3580/3582
Temperature Range98°C
TypeCooling Coil
Unit SizeEach