3,4-Dichlorophenylboronic acid, 97%
3,4-Dichlorophenylboronic acid, 97%
3,4-Dichlorophenylboronic acid, 97%
Acros Organics

3,4-Dichlorophenylboronic acid, 97%

CAS: 151169-75-4 | C6H5BCl2O2 | 190.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 359090250
also known as 35909
HSN 29319090
Price (INR)
39,200.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
39,200.00
Each
Chemical Identifiers
CAS151169-75-4
IUPAC Name(3,4-dichlorophenyl)boronic acid
Molecular FormulaC6H5BCl2O2
InChI KeyJKIGHOARKAIPJI-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(Cl)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Appearance (Form)Powder
Infrared spectrumConforms
Appearance (Color)White to off-white
Titration with NaOH>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only