MaxQ™ 3000/5000 Dedicated Platforms with Clamps
Actual product may vary
MaxQ™ 3000/5000 Dedicated Platforms with Clamps
MaxQ™ 3000/5000 Dedicated Platforms with Clamps
Thermo Scientific™

MaxQ™ 3000/5000 Dedicated Platforms with Clamps

Use these dedicated platforms with Thermo Scientific™ MaxQ™ 3000 and 5000 Shakers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Product TypeLength (English) PlatformNo. of Positions
3599-55MaxQ 3000 Shaker OnlyErlenmeyer Flask36 in.70
3599-46MaxQ 3000, MaxQ 5000 Shakers OnlyUtility Tray30 in.1
3599-5MaxQ 3000, MaxQ 5000 Shakers OnlyErlenmeyer Flask30 in.40
3599-58MaxQ 3000 Shaker OnlyErlenmeyer Flask36 in.15
3599-63MaxQ 3000, MaxQ 5000 Shakers OnlyAdjustable Vessel Platform30 in.Adjustable
3599-96MaxQ 3000 ShakersSeparatory Funnel36 in.16
Catalog number 3599-55
Price (INR)
-
For Use With (Equipment):
MaxQ 3000 Shaker Only
Product Type:
Erlenmeyer Flask
Length (English) Platform:
36 in.
No. of Positions:
70

Achieve maximum capacity if only using a single vessel size such as a flask with Thermo Scientific™ MaxQ™ 3000/5000 Dedicated Platforms with Clamps. Use these dedicated platforms with MaxQ™ 3000 and 5000 Shakers.

  • Clamps come installed to provide maximum capacity
Specifications
Dimensions (L x W)36 x 24 in.
For Use With (Equipment)MaxQ 3000 Shaker Only
Holds300 mL
Length (English) Platform36 in.
Length (Metric) Platform91 cm
No. of Positions70
Product TypeErlenmeyer Flask
Width (English) Platform24 in.
Width (Metric) Platform61 cm
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
Unit SizeEach