See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
Home
▸
Shop All Products
▸
Chromatography Supplies
▸
Chromatography Ferrule and Nut Kits
▸
SLIPFREE Ferrules
Actual product may vary
Thermo Scientific™
SLIPFREE Ferrules
Thermo Scientific™ SLIPFREE Ferrules are for use with SLIPFREE connectors for HPLC columns.
Have Questions?
Catalog Number
Material
36024
Vespel
Catalog number
36024
Price (INR)
-
Material:
Vespel
Thermo Scientific™ SLIPFREE Ferrules are for use with SLIPFREE connectors for HPLC columns.
Vespel ferrules replace the standard Vespel ferrules supplied with SLIPFREE column connectors
Kel-F and PEEK are offered for applications in which Vespel is not suitable
Specifications
Description
Vespel sealing ferrule
For Use With (Equipment)
SLIPFREE Connectors for HPLC Columns
Material
Vespel
Type
Sealing Ferrule