2,5-Diaminopyridine, 97%
2,5-Diaminopyridine, 97%
2,5-Diaminopyridine, 97%
Acros Organics

2,5-Diaminopyridine, 97%

CAS: 4318-76-7 | C5H7N3 | 109.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number 360690050
also known as 36069
HSN 29333919
Price (INR)
32,619.00
Each
Quantity:
5 g
Request bulk or custom format
Price (INR)
32,619.00
Each
Chemical Identifiers
CAS4318-76-7
IUPAC Namepyridine-2,5-diamine
Molecular FormulaC5H7N3
InChI KeyMIROPXUFDXCYLG-UHFFFAOYSA-N
SMILESNC1=CC=C(N)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red-brown to brown to purple to black
Appearance (Form)Powder and/or chunks
Infrared spectrumConforms
HPLC>=96.0 %
Titration with HClO496.0 to 104.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only