Nalgene™ Natural HDPE Diagnostic Bottles without Closure: Bulk Pack
Actual product may vary

Nalgene™ Natural HDPE Diagnostic Bottles without Closure: Bulk Pack

Store and ship small volume, valuable reagents with natural, translucent HDPE diagnostic bottles, which are packed without closures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Neck Finish
362002-90258 mL20-415 mm
362002-91254 mL13-415 mm
362002-9050
also known as 362002-9050EP
15 mL20-415 mm
Catalog number 362002-9025
HSN 39233090
Price (INR)
76,303.00
Case
Capacity (Metric):
8 mL
Neck Finish:
20-415 mm
Price (INR)
76,303.00
Case

Store and ship small volume, valuable reagents with Thermo Scientific™ Nalgene™ Natural HDPE Diagnostic Bottles without Closure, for use in test kits and small-volume applications. These high-density polyethylene bottles are packed without closures so they can be matched with various colored closures (Cat. No. 362150). With uniform walls resistant to splitting or puncturing, these high-density polyethylene bottles are leakproof† when assembled with Nalgene closures (ordered separately).

Features:
  • Nonsterile
  • Natural, translucent high-density polyethylene
  • Durable, uniform walls resistant to splitting or puncturing

Ordering Information:

Bottles are bulk packed without closures. Packed 2000 per case. Order colored closures separately (Cat. No. 362150).

  • A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
Capacity0.25 oz., 8 mL
Capacity (Metric)8 mL
Capacity (English)0.25 oz.
ColorTranslucent
Description8mL, 20-415 neck finish, bulk pack
Diameter (Metric) Inner Neck13 mm
MaterialHDPE
No. per Case2000
No. per Pack2000
SterilityNon-sterile
Neck Finish20-415 mm
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Diagnostic Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure IncludedNo
MouthNarrow
Height (Metric)44 mm
Product TypeSample Container
Width (English)0.98 in.
Height (English)1.73 in.
Diameter (Metric)25 mm
Through the Closure DispensingNo
Unit SizeCase