Nalgene™ HDPE Diagnostic Bottles without Closure: Bulk Pack
Nalgene™ HDPE Diagnostic Bottles without Closure: Bulk Pack
Actual product may vary
Nalgene™ HDPE Diagnostic Bottles without Closure: Bulk Pack
Nalgene™ HDPE Diagnostic Bottles without Closure: Bulk Pack
Nalgene™ HDPE Diagnostic Bottles without Closure: Bulk Pack

Nalgene™ HDPE Diagnostic Bottles without Closure: Bulk Pack

Store samples and ship small volumes of valuable reagents and light-sensitive diagnostic samples with these HDPE diagnostic bottles without closure.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Neck Finish
362008-91254 mL13-415 mm
362008-90258 mL20-415 mm
362008-905015 mL20-415 mm
Catalog number 362008-9125
HSN 39233090
Price (INR)
87,252.00
Case
Add to cart
Capacity (Metric):
4 mL
Neck Finish:
13-415 mm
Price (INR)
87,252.00
Case
Add to cart

Store samples and ship small volumes of valuable reagents and light-sensitive diagnostic samples with Thermo Scientific™ Nalgene™ HDPE Diagnostic Bottles without Closure. Safe and leakproof† when used with Nalgene closures, these bottles can also be used in test kits and many other small-volume applications. Bottles are made of opaque white, high-density polyethylene and are packed without closures. They can be matched with various colored closures (Cat. No. 362150 series).

Features:
  • Nonsterile
  • Durable, uniform walls are resistant to splitting or puncturing
  • Leakproof† when assembled with Nalgene closure (must be ordered separately)

Ordering Information:

Bottles are bulk packed without closures. 2000 per case. Order colored closures separately (Cat. No. 362150).

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
Capacity0.125 oz., 4 mL
Capacity (Metric)4 mL
Capacity (English)0.125 oz.
ColorOpaque White
Diameter (Metric) Inner Neck8 mm
MaterialHDPE
No. per Case2000
No. per Pack2000
SterilityNon-sterile
Neck Finish13-415 mm
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Classs VI. Product meets the requirements for light-resistant containers per USP latest edition.
Closure IncludedNo
GraduatedNo
MouthNarrow
ShapeRound
Height (Metric)41 mm
Product TypeSample Container
Width (English)0.62 in.
Height (English)1.61 in.
Closure MaterialPolypropylene
Closure Size13-415 mm
Diameter (Metric)16 mm
Unit SizeCase