Nalgene™ HDPE High-Profile Closures with Color Coders for Micro Packaging Vials: Nonsterile
Nalgene™ HDPE High-Profile Closures with Color Coders for Micro Packaging Vials: Nonsterile
Actual product may vary
Nalgene™ HDPE High-Profile Closures with Color Coders for Micro Packaging Vials: Nonsterile
Nalgene™ HDPE High-Profile Closures with Color Coders for Micro Packaging Vials: Nonsterile
Nalgene™ HDPE High-Profile Closures with Color Coders for Micro Packaging Vials: Nonsterile
Nalgene™ HDPE High-Profile Closures with Color Coders for Micro Packaging Vials: Nonsterile
Enable rapid identification by using these high profile closures with color coders. Offered in a variety of colors, closures have no O-ring to fall out or contaminate contents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
362830-0117Gold
362830-0110Clear
362830-0111White
362830-0112Yellow
362830-0113Orange
362830-0114Green
362830-0115Red
362830-0116Blue
362830-0118Purple
362830-0119Natural
362830-1110Teal
362830-5110Light Tan
362830-5114Light Green
362830-5116Light Blue
362830-5118Light Purple
Catalog number 362830-0117
HSN 39235090
Price (INR)
32,341.00
Case
Add to cart
Color:
Gold
Price (INR)
32,341.00
Case
Add to cart
Enable rapid identification by using Thermo Scientific™ Nalgene™ HDPE High Profile Closures with Color Coders for Micro Packaging Vials, offered in a variety of colors. Closures have no O-ring to fall out or contaminate contents. Color-coded inserts are free of toxic heavy metals and do not come in contact with vial contents. Vials and closures are pressure-tested together at 7.5psig.
Features:
  • High Density Polyethylene
  • Threaded, screw closure
  • Noncytotoxic, nonpyrogenic
  • Nonsterile
  • Leakproof† when used with Nalgene Micro Packaging Vials
Ordering Information:
Micro Packaging Vial Closures are bulk packed without vials, 500 per inner bag, 1000 per case. Order nonsterile vials separately (Cat. Nos. 362800, 362805).
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database
Specifications
DescriptionHigh-profile closures w/color coders for micro packaging vials
ColorGold
Diameter (Metric) Outer11 mm
MaterialHDPE
SterilityNon-sterile
No. per Case1000
No. per Pack500
Product LineNalgene
Closure MaterialHDPE
Closure TypeScrew Cap
Quantity500 Bag, 1000 Cs.
Closure Size11 mm
Certifications/ComplianceISO 13485:2016
Product TypeClosure
Unit SizeCase