(R)-2-Isopropylsuccinic acid-1-methyl ester, 95%, (98% ee), Thermo Scientific Chemicals
(R)-2-Isopropylsuccinic acid-1-methyl ester, 95%, (98% ee), Thermo Scientific Chemicals
(R)-2-Isopropylsuccinic acid-1-methyl ester, 95%, (98% ee), Thermo Scientific Chemicals
Acros Organics

(R)-2-Isopropylsuccinic acid-1-methyl ester, 95%, (98% ee), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 mg
Packaging:
View all
Glass Bottle
Catalog number 363060050
also known as 36306
HSN 29171990
Price (INR)
166,908.00
Each
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
166,908.00
Each
Chemical Identifiers
CAS220498-08-8
IUPAC Name(3R)-3-(methoxycarbonyl)-4-methylpentanoate
Molecular FormulaC8H13O4
InChI KeyVHOGSWBAPDPWTG-ZCFIWIBFSA-M
SMILESCOC(=O)[C@H](CC([O-])=O)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Viscous liquid
Appearance (Color)Clear yellow
Infrared spectrumConforms
GC>=94 %
Enantiomeric excess>=97.5 % (GC)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only