Bis(tri-tert-butylphosphine)palladium(0), 98%
Bis(tri-tert-butylphosphine)palladium(0), 98%
Bis(tri-tert-butylphosphine)palladium(0), 98%
Acros Organics

Bis(tri-tert-butylphosphine)palladium(0), 98%

CAS: 53199-31-8 | C24H54P2Pd | 511.06 g/mol
Have Questions?
Catalog NumberQuantityPackaging
363500010
also known as 36350
1 gGlass bottle
Catalog number 363500010
also known as 36350
HSN 29319090
Price (INR)
51,675.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
51,675.00
Each
Chemical Identifiers
CAS53199-31-8
IUPAC Namebis(tri-tert-butylphosphane) palladium
Molecular FormulaC24H54P2Pd
InChI KeyMXQOYLRVSVOCQT-UHFFFAOYSA-N
SMILES[Pd].CC(C)(C)P(C(C)(C)C)C(C)(C)C.CC(C)(C)P(C(C)(C)C)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to orange-brown
Appearance (Form)Powder or crystals
Assay>=97.5 % (ex Pd)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only