Toluene, 99.85%, Extra Dry over Molecular Sieve, AcroSeal™
Toluene, 99.85%, Extra Dry over Molecular Sieve, AcroSeal™
Toluene, 99.85%, Extra Dry over Molecular Sieve, AcroSeal™
Acros Organics

Toluene, 99.85%, Extra Dry over Molecular Sieve, AcroSeal™

CAS: 108-88-3 | C7H8 | 92.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
500 mL
2.5 L
Catalog number 364410010
HSN 29023000
Price (INR)
8,329.00
Each
Quantity:
1 L
Request bulk or custom format
Price (INR)
8,329.00
Each
Chemical Identifiers
CAS108-88-3
IUPAC Nametoluene
Molecular FormulaC7H8
InChI KeyYXFVVABEGXRONW-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.005 % (Coulometric)
Appearance (Color)Clear colorless
GC>=99.8 %
Acidity (HCl)=<0.002 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only