(1R,2R)-(-)-1,2-Diaminocyclohexane, 99%
(1R,2R)-(-)-1,2-Diaminocyclohexane, 99%
(1R,2R)-(-)-1,2-Diaminocyclohexane, 99%
Acros Organics

(1R,2R)-(-)-1,2-Diaminocyclohexane, 99%

CAS: 20439-47-8 | C6H14N2 | 114.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 364640050
also known as 36464
HSN 29213010
Price (INR)
27,767.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
27,767.00
Each
Chemical Identifiers
CAS20439-47-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Low melting crystalline solid
GC>=98.5 %
Melting point40°C to 45°C
Specific optical rotation-24.5° to -26.5° (20°C, 589 nm) (c=5, 1 N HCl)
Appearance (Color)White to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only