6-Bromo-1-hexene, 97%
6-Bromo-1-hexene, 97%
6-Bromo-1-hexene, 97%
Acros Organics

6-Bromo-1-hexene, 97%

CAS: 2695-47-8 | C6H11Br | 163.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number 364700250
also known as 36470
HSN 29033919
Price (INR)
66,162.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
66,162.00
Each
Chemical Identifiers
CAS2695-47-8
IUPAC Name6-bromohex-1-ene
Molecular FormulaC6H11Br
InChI KeyRIMXEJYJXDBLIE-UHFFFAOYSA-N
SMILESBrCCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Refractive index1.4640 to 1.4670 (20°C, 589 nm)
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only