Coconut oil, pure, refined
Acros Organics

Coconut oil, pure, refined

CAS: 8001-31-8
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 365471000
HSN 98020000
Price (INR)
2,912.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS8001-31-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to light yellow
Appearance (Form)Oily liquid or solid
Acid value=<0.5 mg KOH/g
Iodine value7 to 12 g I2/100 g
Peroxide value=<5 meq O2/kg
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only