Methyl 4-bromobutyrate, 98+%
Methyl 4-bromobutyrate, 98+%
Methyl 4-bromobutyrate, 98+%
Acros Organics

Methyl 4-bromobutyrate, 98+%

CAS: 4897-84-1 | C5H9BrO2 | 181.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number 365830500
also known as 36583
HSN 29159090
Price (INR)
6,841.00
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
6,841.00
Each
Chemical Identifiers
CAS4897-84-1
IUPAC Namemethyl 4-bromobutanoate
Molecular FormulaC5H9BrO2
InChI KeyQAWFLJGZSZIZHO-UHFFFAOYSA-N
SMILESCOC(=O)CCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4610 to 1.4630 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only