1-(2-Methoxyethyl)piperazine, 98%
1-(2-Methoxyethyl)piperazine, 98%
1-(2-Methoxyethyl)piperazine, 98%
Acros Organics

1-(2-Methoxyethyl)piperazine, 98%

CAS: 13484-40-7 | C7H16N2O | 144.21 g/mol
Have Questions?
Catalog NumberQuantityPackaging
367110010
also known as 36711
1 gGlass bottle
Catalog number 367110010
also known as 36711
HSN 29335990
Price (INR)
8,381.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
8,381.00
Each
Chemical Identifiers
CAS13484-40-7
IUPAC Name4-(2-methoxyethyl)piperazine-1,4-diium
Molecular FormulaC7H18N2O
InChI KeyBMEMBBFDTYHTLH-UHFFFAOYSA-P
SMILESCOCC[NH+]1CC[NH2+]CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)Clear colorless to yellow
Refractive index1.4720 to 1.4750 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only