2-Bromoethyl ethyl ether, 90%
2-Bromoethyl ethyl ether, 90%
2-Bromoethyl ethyl ether, 90%
2-Bromoethyl ethyl ether, 90%
Acros Organics

2-Bromoethyl ethyl ether, 90%

CAS: 592-55-2 | C4H9BrO | 153.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
View all
Glass bottle
Catalog number 367400250
also known as 36740
HSN 29091900
Price (INR)
12,049.00
Each
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
12,049.00
Each
Add to cart
Chemical Identifiers
CAS592-55-2
IUPAC Name1-bromo-2-ethoxyethane
Molecular FormulaC4H9BrO
InChI KeyMMYKTRPLXXWLBC-UHFFFAOYSA-N
SMILESCCOCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=88 %
Appearance (Color)Clear colorless to brown
Refractive index1.4435 to 1.4515 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only