Sodium selenate, 98%
Sodium selenate, 98%
Sodium selenate, 98%
Acros Organics

Sodium selenate, 98%

CAS: 13410-01-0 | Na2O4Se | 188.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 369960250
HSN 28429010
Price (INR)
12,546.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
12,546.00
Each
Chemical Identifiers
CAS13410-01-0
SpecificationsSpecification SheetSpecification Sheet
Identificationconfirmed
Appearance (Form)Crystalline powder
Assay>=97.5 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only