1-Methyl-4-iodo-1H-pyrazole, 97%
1-Methyl-4-iodo-1H-pyrazole, 97%
1-Methyl-4-iodo-1H-pyrazole, 97%
Acros Organics

1-Methyl-4-iodo-1H-pyrazole, 97%

CAS: 39806-90-1 | C4H5IN2 | 208 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 371790050
also known as 37179
HSN 29319090
Price (INR)
37,188.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
37,188.00
Each
Chemical Identifiers
CAS39806-90-1
IUPAC Name4-iodo-1-methyl-1H-pyrazole
Molecular FormulaC4H5IN2
InChI KeyRSDRDHPLXWMTRJ-UHFFFAOYSA-N
SMILESCN1C=C(I)C=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Semitransparent crystals or crystalline needles
GC>=96.0 %
Appearance (Color)Off-white
Infrared spectrumConforms
Melting point61°C to 67°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only