4,4'-Dimethyl-2,2'-bipyridyl, 99+%
4,4'-Dimethyl-2,2'-bipyridyl, 99+%
4,4'-Dimethyl-2,2'-bipyridyl, 99+%
Acros Organics

4,4'-Dimethyl-2,2'-bipyridyl, 99+%

CAS: 1134-35-6 | C12H12N2 | 184.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number 373230050
HSN 29333990
Price (INR)
14,440.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
14,440.00
Each
Chemical Identifiers
CAS1134-35-6
IUPAC Name4,4'-dimethyl-2,2'-bipyridine
Molecular FormulaC12H12N2
InChI KeyNBPGPQJFYXNFKN-UHFFFAOYSA-N
SMILESCC1=CC=NC(=C1)C1=CC(C)=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige
Appearance (Form)Crystalline powder
GC>=99.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only