4-Mercaptobenzoic acid, 90%
4-Mercaptobenzoic acid, 90%
4-Mercaptobenzoic acid, 90%
Acros Organics

4-Mercaptobenzoic acid, 90%

CAS: 1074-36-8 | C7H6O2S | 154.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 373300250
also known as 37330
HSN 29309099
Price (INR)
47,059.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
47,059.00
Each
Chemical Identifiers
CAS1074-36-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Melting point215°C to 224°C
HPLC>=88 %
Appearance (Color)White to brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only