SoftFit-L™ Non-Filtered Low Retention Hinged Rack Pipette Tips

SoftFit-L™ Non-Filtered Low Retention Hinged Rack Pipette Tips

Pipette samples with the accuracy and precision you expect from your pipette, every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Sterility
3751-05-HRPK
also known as 3751-05-HR
200 μLSterile
3721-05-HRPK
also known as 3721-05-HR
20 μLSterile
3781-05-HRPK
also known as 3781-05-HR
1000 μLSterile
3782-05-HRPK
also known as 3782-05-HR
1000 μLNon-sterile
Catalog number 3751-05-HRPK
also known as 3751-05-HR
Price (INR)
-
Volume (Metric):
200 μL
Sterility:
Sterile
Pipette samples with the accuracy and precision you expect from your pipette, every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
  • Low-retention tips maximize sample delivery
  • Hinged rack provides easy one-hand access to tips
  • Color-coded inserts allows quick identification of tip volume
Specifications
Clean ClaimsDNase, RNase, DNA, ATP, and Endotoxin Free
Description200L, Non-Filtered, Sterile, Hinged Rack
Length (English)1.97 in.
Length (Metric)5.00cm
Model200L
ColorNatural
Color CodeGreen
FilteredNon-filtered
For Use With (Equipment)Rainin LTS LiteTouch
FormatHinged Rack
Product LineART SoftFit L
SterilitySterile
Tip StyleLTS-style tip with positive stop
Volume (Metric)200 μL
Unit SizePack of 960