SoftFit-L™ Pipette Tips in Hinged Racks

SoftFit-L™ Pipette Tips in Hinged Racks

Gain the accuracy and precision you expect from your pipette samples every time with these pipette tips.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Sterility
3751-HRPK
also known as 3751-HR
200 μLSterile
3721-HRPK
also known as 3721-HR
20 μLSterile
3781-HRPK
also known as 3781-HR
1000 μLSterile
3782-HRPK
also known as 3782-05-HR
1000 μLNon-sterile
3752-HRPK
also known as R-HR-HR
200 μLNon-sterile
3732-HRPK
also known as 3732-HR
300 μLNon-sterile
3731-HRPK
also known as 3731-HR
300 μLSterile
3722-HRPK
also known as 3722-HR
20 μLNon-sterile
3741-HRPK
also known as 3741-HR
1200 μLSterile
Catalog number 3751-HRPK
also known as 3751-HR
Price (INR)
-
Volume (Metric):
200 μL
Sterility:
Sterile
Gain the accuracy and precision you expect from your pipette samples every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
  • Hinged rack provides easy one-hand access to tips
  • Color-coded inserts allow quick identification of tip volume
Specifications
Clean ClaimsDNase, RNase, DNA, ATP, and Endotoxin Free
Description200L, Non-Filtered, Sterile, Hinged Rack
Length (English)1.97 in.
Length (Metric)5.00cm
Model200L
ColorNatural
Color CodeGreen
FilteredNon-filtered
For Use With (Equipment)Rainin LTS LiteTouch
FormatHinged Rack
Product LineART SoftFit L
SterilitySterile
Tip StyleUniversal
Volume (Metric)200 μL
Unit SizePack of 960