Search
Pipette samples with the accuracy and precision you expect from your pipette, every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
| Catalog Number | Volume (Metric) | Sterility | Unit Size |
|---|---|---|---|
| 3752-RIPK also known as 3752-RI | 200 μL | Non-sterile | Pack of 960 |
| 3722-RIPK also known as 3722-RI | 20 μL | Non-sterile | Pack of 960 |
| 3732-RIPK also known as 3732-RI | 300 μL | Non-sterile | Pack of 960 |
| 3742-RI | 1200 μL | Non-sterile | Case of 3072 |
| 3742-RIPK also known as 3742-RI | 1200 μL | Non-sterile | Pack of 768 |
| 3782-RIPK also known as 3782-RI | 1000 μL | Non-sterile | Pack of 768 |
| 3721-RIPK also known as 3721-RI | 20 μL | Sterile | Pack of 960 |
| 3731-RIPK also known as 3731-RI | 300 μL | Sterile | Pack of 960 |
| 3741-RI | 1200 μL | Sterile | Case of 3072 |
| 3741-RIPK also known as 3741-RI | 1200 μL | Sterile | Pack of 768 |
| 3751-RIPK also known as 3751-RI | 200 μL | Sterile | Pack of 960 |
| 3781-RIPK also known as 3781-RI | 1000 μL | Sterile | Pack of 768 |
The SoftFit-L Reload System is a complete product offering of pipet tips compatible with LTS™ pipetters made by Rainin Instrument, LLC. Designed in response to customer demands, this unique system provides preloaded pipet tips in color-coded inserts by volume size with the option of an easy-to-use 'push down' reload pack. Choose sterile or nonsterile tips, in five volume sizes, with or without our proprietary low-retention technology.