3-Phenyl-1-propyne, 97%, stabilized
3-Phenyl-1-propyne, 97%, stabilized
3-Phenyl-1-propyne, 97%, stabilized
Acros Organics

3-Phenyl-1-propyne, 97%, stabilized

CAS: 10147-11-2 | C9H8 | 116.16 g/mol
Have Questions?
Catalog NumberQuantityPackaging
376800050
also known as 37680
5 gGlass bottle
Catalog number 376800050
also known as 37680
HSN 29029090
Price (INR)
27,956.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
27,956.00
Each
Chemical Identifiers
CAS10147-11-2
IUPAC Name(prop-2-yn-1-yl)benzene
Molecular FormulaC9H8
InChI KeyNGKSKVYWPINGLI-UHFFFAOYSA-N
SMILESC#CCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
GC>=96.0 %
Infrared spectrumConforms
Stabilizer100 to 400 ppm BHT
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only