(Trimethylsilyl)methylmagnesium chloride, 1.3M solution in THF, AcroSeal™
(Trimethylsilyl)methylmagnesium chloride, 1.3M solution in THF, AcroSeal™
(Trimethylsilyl)methylmagnesium chloride, 1.3M solution in THF, AcroSeal™
Acros Organics

(Trimethylsilyl)methylmagnesium chloride, 1.3M solution in THF, AcroSeal™

CAS: 13170-43-9 | C4H11ClMgSi | 146.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
100 mL
Packaging:
AcroSeal™ Glass bottle
Catalog number 377468000
also known as 37746
HSN 29319090
Price (INR)
126,051.00
Each
Quantity:
800 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (INR)
126,051.00
Each
Chemical Identifiers
CAS13170-43-9
IUPAC Name[(chloromagnesio)methyl]trimethylsilane
Molecular FormulaC4H11ClMgSi
InChI KeyNAQATMJWCJCHOZ-UHFFFAOYSA-M
SMILESC[Si](C)(C)C[Mg]Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Clear to hazy grey to brown-black
Titration Grignard1.2 to 1.5 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only