SoftFit-L™ Non-Filtered Low Retention Hinged Rack Pipette Tips

SoftFit-L™ Non-Filtered Low Retention Hinged Rack Pipette Tips

Pipette samples with the accuracy and precision you expect from your pipette, every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Sterility
3782-05-HRPK
also known as 3782-05-HR
1000 μLNon-sterile
3721-05-HRPK
also known as 3721-05-HR
20 μLSterile
3751-05-HRPK
also known as 3751-05-HR
200 μLSterile
3781-05-HRPK
also known as 3781-05-HR
1000 μLSterile
Catalog number 3782-05-HRPK
also known as 3782-05-HR
Price (INR)
-
Volume (Metric):
1000 μL
Sterility:
Non-sterile
Pipette samples with the accuracy and precision you expect from your pipette, every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
  • Low-retention tips maximize sample delivery
  • Hinged rack provides easy one-hand access to tips
  • Color-coded inserts allows quick identification of tip volume
Specifications
Clean ClaimsDNase, RNase, DNA, ATP, and Endotoxin Free
Description1000L, Non-Filtered, Non-sterile, Hinged Rack
Length (English)3.39 in.
Length (Metric)8.61cm
Model1000L
ColorNatural
Color CodeBlue
FilteredNon-filtered
For Use With (Equipment)Rainin LTS LiteTouch
FormatHinged Rack
Product LineART SoftFit L
SterilityNon-sterile
Tip StyleLTS-style tip with positive stop
Volume (Metric)1000 μL
Unit SizePack of 768