SoftFit-L™ Pipette Tips, TipCycle Recycle Kit, Hinged Racks
Thermo Scientific™
SoftFit-L™ Pipette Tips, TipCycle Recycle Kit, Hinged Racks
The pipette tip and rack mail-back recycling program provides a simple and effective solution for recycling single use plastics. The TipCycle Recycle Kit arrives with your case of pipette tips to collect used pipette tips and empty racks. Attach the pre-paid shipping label and return for recycling.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Sterility
3782RC-HR1000 μLNon-sterile
3721RC-HR20 μLSterile
3722RC-HR20 μLNon-sterile
3731RC-HR300 μLSterile
3732RC-HR300 μLNon-sterile
3741RC-HR1200 μLSterile
3742RC-HR1200 μLNon-sterile
3751RC-HR200 μLSterile
3752RC-HR200 μLNon-sterile
3781RC-HR1000 μLSterile
Catalog number 3782RC-HR
Price (INR)
-
Volume (Metric):
1000 μL
Sterility:
Non-sterile
Gain the accuracy and precision you expect from your pipette samples every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
  • Recycle nonhazardous pipette tips, racks, and associated packaging with Tipcycle
  • Complete TipCycle Pipette Tip Recycling Kit Includes: 1 case of pipette tips, box liner, liner tie, FedEx return shipping label, waiver form, and instructions for return
  • Hinged rack provides easy one-hand access to tips
  • Color-coded inserts allow quick identification of tip volume
Specifications
Clean ClaimsDNase, RNase, DNA, Pryogen Free
Includes1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
Length (English)3.39 in.
Length (Metric)8.61 cm
Model1000 μL, Non-Filtered, Non-sterile, Hinged Rack, TipCycle Recycle Kit
Product TypeTipCycle, Pipette Tip and Rack Recycling Kit
AutoclavableNot Autoclavable
ColorNatural
Color CodeBlue
FilteredNon-filtered
FormatHinged Rack
Product LineSoftfit-L
SterilityNon-sterile
Tip StyleSpecialty
Volume (Metric)1000 μL
Unit SizeCase of 3072