4-Ethylbenzaldehyde, 98%
4-Ethylbenzaldehyde, 98%
4-Ethylbenzaldehyde, 98%
Acros Organics

4-Ethylbenzaldehyde, 98%

CAS: 4748-78-1 | C9H10O | 134.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 378950250
HSN 29122990
Price (INR)
7,504.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
7,504.00
Each
Chemical Identifiers
CAS4748-78-1
IUPAC Name4-ethylbenzaldehyde
Molecular FormulaC9H10O
InChI KeyQNGNSVIICDLXHT-UHFFFAOYSA-N
SMILESCCC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)Clear colorless to light yellow
Refractive index1.5385 to 1.5405 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only