Matrix™ Open-Top Tubes in Barcoded Latch Racks
Matrix™ Open-Top Tubes in Barcoded Latch Racks
Matrix™ Open-Top Tubes in Barcoded Latch Racks
Matrix™ Open-Top Tubes in Barcoded Latch Racks
Matrix™ Open-Top Tubes in Barcoded Latch Racks

Matrix™ Open-Top Tubes in Barcoded Latch Racks

Securely store and track samples in Thermo Scientific™ Matrix™ Open-Top tubes. Laser etching produces a permanent, high-contrast, 2D barcode on each tube, offering maximum reliability and traceability for chemical, biological, and genomic samples.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberBottom ShapeSterility
3792-BRV-Bottom WellSterile
3712-11-BRFlat WellNon-sterile
3791-BRV-Bottom WellNon-sterile
Catalog number 3792-BR
Price (INR)
-
Request A Quote
Bottom Shape:
V-Bottom Well
Sterility:
Sterile
Securely store and track samples in Thermo Scientific™ Matrix™ Open-Top tubes. Laser etching produces a permanent, high-contrast, 2D barcode on each tube, offering maximum reliability and traceability for chemical, biological, and genomic samples. Barcoded latch racks feature a linear barcode, 2D barcode, and human readable rack identification on three sides of the rack. Use in combination with Thermo Scientific™ Matrix™ SepraSeal and DurSeal capping systems and the Thermo Scientific™ VisionMate™ family of barcode readers.

Secure Tracking

  • A unique 2D barcode is laser-etched onto the base of every polypropylene open-top tube to securely identify and track samples
  • 2D barcodes are resistant to 100% DMSO, ideal for compound processing and storage
  • Non-proprietary 2D codes can be scanned with the Thermo Scientific VisionMate family of barcode readers

Superior Storage Format

  • ANSI-format latch racks maximize storage space and are compatible with multichannel manual pipettes or automated liquid handling equipment
  • Barcoded latch racks include 1D and 2D barcodes, plus human readable text on three sides to provide added same traceability
  • The latch rack lid can be lifted robotically for 2D tube access with automated liquid handling systems and high throughput applications

Stringent Quality Control

  • Every 2D barcoded storage tube is scanned to guarantee readability.
  • Each code is checked against a full database of all previously assigned 2D codes to ensure zero duplicates across the entire line of Matrix 2D barcoded storage tubes.

Storage Tracking Solutions for Any Laboratory

  • Custom 2D coding options are available to meet your laboratory's individual database or tracking requirements
  • Tubes are available with a range of sealing options including Thermo Scientific SepraSeals and DuraSeals
  • SepraSeals are available in a range of color options for visual categorization of samples
  • Pre-Split SepraSeals allow for repeated sample access in automation

Recommended for:

Drug discovery, compound storage, secure sample transport, individual sample retrieval, instant sample identification, secure tracking of archived samples

Specifications
BarcodeTube barcode: Non-proprietary, 12x12 Data-matrix with ECC200 Built-in Error Correction; Rack barcode: 2D Datamatrix; Linear EAN Code 128; Human Readable
ColorColorless
Description2D Barcoded Open-Top Tubes in Barcoded Latch Racks
Packaging Quantity10 Latch Racks of 96 tubes per Case
Product LineMatrix
AutoclavableNot Autoclavable
Bottom ShapeV-Bottom Well
Capacity (Metric)1.4 mL
GraduatedNo
MaterialPolypropylene
Product TypeOpen-top Tube
SterilitySterile
Unit SizeCase of 960