ART™ Non-Filtered Extended Length Hinged Rack Pipette Tips
Thermo Scientific™
ART™ Non-Filtered Extended Length Hinged Rack Pipette Tips
Access tips one-handed with this versatile hinged rack, designed for the way you work.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityTip StyleFit for Purpose
3792-HRPK
also known as 3792-HR
1000 μLNon-sterileUniversal FitCertified Quality
3512-HRPK10 μLNon-sterileUniversal FitCertified Quality
3542-HRPK
also known as 3542-HR
200 μLNon-sterileUniversal FitCertified Quality
3541-HRPK
also known as 3541-HR
200 μLSterileUniversal FitSterile
3542-05-HRPK
also known as 3542-05-HR
200 μLNon-sterileLow RetentionCertified Quality
3541-05-HRPK
also known as 3541-05-HR
200 μLSterileLow RetentionSterile
3792-05-HRPK
also known as 3792-05-HR
1000 μLNon-sterileLow RetentionCertified Quality
3791-HRPK
also known as 3791-HR
1000 μLSterileUniversal FitSterile
3791-05-HRPK
also known as 3791-05-HR
1000 μLSterileLow RetentionSterile
3151-HRPK
also known as 3151-HR
1250 μLSterileUniversal FitSterile
Catalog number 3792-HRPK
also known as 3792-HR
Price (INR)
5,398.00
Pack of 768
Volume (Metric):
1000 μL
Sterility:
Non-sterile
Tip Style:
Universal Fit
Fit for Purpose:
Certified Quality
Price (INR)
5,398.00
Pack of 768
Access tips with one hand using Thermo Scientific™ ART™ Non-Filtered Extended Length Hinged Rack Pipette Tips, designed for the way you work. No longer worry about your tips spilling on your bench or the floor. Maximize your productivity and easily transition from hinged to lift-off in just three easy steps. Simply unhinge, turn and realign the cover — now you have a traditional lift-off when you need it.
Features:
  • Complete selection of non-filtered and specialty tips deliver proven ART quality, performance and reliability, backed by the highest Thermo Scientific standards and worldwide support
  • DNase, RNase, DNA, ATP, and Endotoxin Free
  • Racks offer quick and easy access to pipette tips, saving valuable time
  • Front latch closes and locks, ensuring safe and secure storage throughout
Specifications
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
DescriptionART Non-Filtered, Non-sterile, Rack
Length (English)4.7 in.
Length (Metric)11.85cm
ModelART Non-filtered
ColorNatural
FilteredNon-filtered
Fit for PurposeCertified Quality
For Use With (Equipment)Universal Fit
FormatRack
Product LineART
SterilityNon-sterile
Tip StyleUniversal Fit
Volume (Metric)1000 μL
Unit SizePack of 768