Potassium hydroxide, pure, 8N solution in water
Potassium hydroxide, pure, 8N solution in water
Potassium hydroxide, pure, 8N solution in water
Acros Organics

Potassium hydroxide, pure, 8N solution in water

CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 L
500 mL
Packaging:
Plastic bottle
Catalog number 380620020
also known as 38062
HSN 28152000
Price (INR)
8,664.00
Each
Quantity:
2 L
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1310-58-3
IUPAC Namepotassium hydroxide
Molecular FormulaHKO
InChI KeyKWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES[OH-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Strength7.984 to 8.016 N (20°C)
Appearance (Color)Clear colorless
Appearance (Form)Solution
Composition448 g/l (g KOH/l H20)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only