Methyl 3-nitrosalicylate, 98%, Thermo Scientific Chemicals
Methyl 3-nitrosalicylate, 98%, Thermo Scientific Chemicals
Methyl 3-nitrosalicylate, 98%, Thermo Scientific Chemicals
Acros Organics

Methyl 3-nitrosalicylate, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
3819200505 gGlass bottle
Catalog number 381920050
HSN 29183090
Price (INR)
25,399.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
25,399.00
Each
Chemical Identifiers
CAS22621-41-6
IUPAC Name2-(methoxycarbonyl)-6-nitrobenzen-1-olate
Molecular FormulaC8H6NO5
InChI KeyNIBVYEHAFBEVFI-UHFFFAOYSA-M
SMILESCOC(=O)C1=CC=CC(=C1[O-])[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point127°C to 132°C
GC>=97.5 %
Appearance (Color)Yellow to beige to grey to brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only