Dimethylanilinium Tetrakis (pentafluorophenyl)borate, 98%
Dimethylanilinium Tetrakis (pentafluorophenyl)borate, 98%
Dimethylanilinium Tetrakis (pentafluorophenyl)borate, 98%
Acros Organics

Dimethylanilinium Tetrakis (pentafluorophenyl)borate, 98%

CAS: 118612-00-3 | C32H12BF20N | 801.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 382400010
also known as 38240
HSN 29214290
Price (INR)
34,442.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
34,442.00
Each
Chemical Identifiers
CAS118612-00-3
IUPAC NameN,N-dimethylanilinium; tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
Molecular FormulaC32H12BF20N
InChI KeyBRHZQNMGSKUUMN-UHFFFAOYSA-O
SMILESC[NH+](C)C1=CC=CC=C1.FC1=C(F)C(F)=C(C(F)=C1F)[B-](C1=C(F)C(F)=C(F)C(F)=C1F)(C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
AppearanceMay have a blue hue
Infrared spectrumConforms
NMR (19F-NMR)>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only