4-bromo-2-fluorobenzylamine, 98%
4-bromo-2-fluorobenzylamine, 98%
4-bromo-2-fluorobenzylamine, 98%
Acros Organics

4-bromo-2-fluorobenzylamine, 98%

CAS: 112734-22-2 | C7H7BrFN | 204.04 g/mol
Have Questions?
Catalog NumberQuantityPackaging
384440050
also known as 38444
5 gGlass bottle
Catalog number 384440050
also known as 38444
HSN 29214910
Price (INR)
15,931.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
15,931.00
Each
Chemical Identifiers
CAS112734-22-2
IUPAC Name1-(4-bromo-2-fluorophenyl)methanamine
Molecular FormulaC7H7BrFN
InChI KeyRLTFBWCBGIZCDQ-UHFFFAOYSA-N
SMILESNCC1=C(F)C=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.5630 to 1.5660 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only