Graphite, powder
Graphite, powder
Graphite, powder
Acros Organics

Graphite, powder

CAS: 7782-42-5 | C | 12.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
Packaging:
Glass bottle
Catalog number 385030010
HSN 38011000
Price (INR)
13,043.00
Each
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
13,043.00
Each
Chemical Identifiers
CAS7782-42-5
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfur (S)=<0.05 %
Ash=<0.8 %
Carbon (C)>=99.0 %
Appearance (Color)Black
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
RUO – Research Use Only