Calcium hydroxide, 99.995%, (trace metal basis)
Calcium hydroxide, 99.995%, (trace metal basis)
Calcium hydroxide, 99.995%, (trace metal basis)
Acros Organics

Calcium hydroxide, 99.995%, (trace metal basis)

CAS: 1305-62-0 | CaH2O2 | 74.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3856100505 gGlass bottle
Catalog number 385610050
HSN 28259040
Price (INR)
11,694.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
11,694.00
Each
Chemical Identifiers
CAS1305-62-0
IUPAC Namecalcium dihydroxide
Molecular FormulaCaH2O2
InChI KeyAXCZMVOFGPJBDE-UHFFFAOYSA-L
SMILES[OH-].[OH-].[Ca++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Appearance (Color)White
Total trace metal impurities=<50 ppm
Titration Complexometric>=54 % (Ca)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only