(-)-8-Phenylmenthol, 97%, Thermo Scientific Chemicals
(-)-8-Phenylmenthol, 97%, Thermo Scientific Chemicals
(-)-8-Phenylmenthol, 97%, Thermo Scientific Chemicals
Acros Organics

(-)-8-Phenylmenthol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Packaging:
Glass bottle
Catalog number 387950010
also known as 38795
HSN 29061990
Price (INR)
34,584.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
34,584.00
Each
Chemical Identifiers
CAS65253-04-5
IUPAC Name(1R,2S,5R)-5-methyl-2-(2-phenylpropan-2-yl)cyclohexan-1-ol
Molecular FormulaC16H24O
InChI KeyWTQIZFCJMGWUGZ-BPLDGKMQSA-N
SMILESC[C@@H]1CC[C@H]([C@H](O)C1)C(C)(C)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Viscous liquid
GC>=96.0 %
Refractive index1.5285 to 1.5315 (20°C, 589 nm)
Specific optical rotation-22° to -27° (20°C, 589 nm) (c=1, ethanol)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only