Dimethylamine, 2M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Dimethylamine, 2M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Dimethylamine, 2M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Acros Organics

Dimethylamine, 2M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
100 mL
Catalog number 388388000
also known as 38838
HSN 29211110
Price (INR)
44,195.00
Each
Quantity:
800 mL
Request bulk or custom format
Price (INR)
44,195.00
Each
Chemical Identifiers
CAS124-40-3
IUPAC Namedimethylamine
Molecular FormulaC2H7N
InChI KeyROSDSFDQCJNGOL-UHFFFAOYSA-N
SMILESCNC
View more
SpecificationsSpecification SheetSpecification Sheet
Strength1.8 to 2.2 M
Appearance (Form)Solution
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only