Phosphorus oxybromide, 95%
Phosphorus oxybromide, 95%
Phosphorus oxybromide, 95%
Acros Organics

Phosphorus oxybromide, 95%

CAS: 7789-59-5 | Br3OP | 286.69 g/mol
Have Questions?
Catalog NumberQuantityPackaging
388451000
also known as 38845
100 gGlass bottle
Catalog number 388451000
also known as 38845
HSN 28261990
Price (INR)
35,839.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
35,839.00
Each
Chemical Identifiers
CAS7789-59-5
IUPAC Namephosphoroyl tribromide
Molecular FormulaBr3OP
InChI KeyUXCDUFKZSUBXGM-UHFFFAOYSA-N
SMILESBrP(Br)(Br)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals or fused mass
Appearance (Color)Colorless to yellow to orange to light brown
Titration Argentometric>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only