Zinc(II) acetylacetonate, ca. 25% Zn
Zinc(II) acetylacetonate, ca. 25% Zn
Zinc(II) acetylacetonate, ca. 25% Zn
Acros Organics

Zinc(II) acetylacetonate, ca. 25% Zn

CAS: 14024-63-6 | C10H14O4Zn | 263.60 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
Packaging:
Glass bottle
Catalog number 388771000
also known as 38877
HSN 98020000
Price (INR)
10,131.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
10,131.00
Each
Chemical Identifiers
CAS14024-63-6
IUPAC Namezinc(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC10H14O4Zn
InChI KeyCYDXJXDAFPJUQE-FDGPNNRMSA-L
SMILES[Zn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration Complexometric23.5 to 26.0 % (Zinc)
Appearance (Color)White to off-white
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only