Sodium hydrogen tartrate, 98+%, for analysis
Sodium hydrogen tartrate, 98+%, for analysis
Sodium hydrogen tartrate, 98+%, for analysis
Acros Organics

Sodium hydrogen tartrate, 98+%, for analysis

CAS: 526-94-3 | C4H5NaO6 | 172.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 389422500
HSN 29181390
Price (INR)
6,645.00
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
6,645.00
Each
Chemical Identifiers
CAS526-94-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Insoluble matter=<0.03 % (in water)
Appearance (Color)White
Titration with NaOH>=98.0 %
Chloride (Cl)=<0.02 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only