[Hydroxy(tosyloxy)iodo]benzene, 97%
[Hydroxy(tosyloxy)iodo]benzene, 97%
[Hydroxy(tosyloxy)iodo]benzene, 97%
Acros Organics

[Hydroxy(tosyloxy)iodo]benzene, 97%

CAS: 27126-76-7 | C13H13IO4S | 392.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 389610250
also known as 38961
HSN 29093090
Price (INR)
22,393.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
22,393.00
Each
Chemical Identifiers
CAS27126-76-7
Molecular FormulaC13H13IO4S
InChI KeyLRIUKPUCKCECPT-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)S(=O)(=O)O[I](O)C1=CC=CC=C1
Molecular Weight (g/mol)392.21
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point130°C to 137°C
Titration with NaOH96.0 to 104.0 %
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
oxidation reagent
RUO – Research Use Only