Sodium phosphate, tribasic, pure, anhydrous
Sodium phosphate, tribasic, pure, anhydrous
Sodium phosphate, tribasic, pure, anhydrous
Acros Organics

Sodium phosphate, tribasic, pure, anhydrous

CAS: 7601-54-9 | Na3O4P | 163.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
1 kg
2.5 kg
Packaging:
View all
Fiber Drum
Catalog number 389810100
HSN 28353900
Price (INR)
47,177.00
Each
Quantity:
10 kg
Packaging:
Fiber Drum
Request bulk or custom format
Price (INR)
47,177.00
Each
Chemical Identifiers
CAS7601-54-9
IUPAC Nametrisodium phosphate
Molecular FormulaNa3O4P
InChI KeyRYFMWSXOAZQYPI-UHFFFAOYSA-K
SMILES[Na+].[Na+].[Na+].[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with HCl>=96.0 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only