Calcium phosphate, dibasic dihydrate, 98+%, extra pure
Calcium phosphate, dibasic dihydrate, 98+%, extra pure
Calcium phosphate, dibasic dihydrate, 98+%, extra pure
Acros Organics

Calcium phosphate, dibasic dihydrate, 98+%, extra pure

CAS: 7789-77-7 | CaHO42H2O | 172.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
389830050
also known as 38983
5 kgPlastic drum
Catalog number 389830050
also known as 38983
HSN 28352500
Price (INR)
25,944.00
Each
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS7789-77-7
IUPAC Namecalcium dihydrate hydrogen phosphate
Molecular FormulaCaH5O6P
InChI KeyXAAHAAMILDNBPS-UHFFFAOYSA-L
SMILESO.O.[Ca++].OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder
IdentificationCalcium: positive
IdentificationPhosphate: positive
Loss on ignition24.5 % to 26.5 % (800°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only