Sodium phosphate, monobasic, 99%, for analysis, anhydrous
Sodium phosphate, monobasic, 99%, for analysis, anhydrous
Sodium phosphate, monobasic, 99%, for analysis, anhydrous
Acros Organics

Sodium phosphate, monobasic, 99%, for analysis, anhydrous

CAS: 7558-80-7 | H2NaO4P | 119.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
250 g
1 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 389870025
also known as 38987
HSN 28352200
Price (INR)
19,387.00
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
19,387.00
Each
Chemical Identifiers
CAS7558-80-7
IUPAC Namesodium dihydrogen phosphate
Molecular FormulaH2NaO4P
InChI KeyAJPJDKMHJJGVTQ-UHFFFAOYSA-M
SMILES[Na+].OP(O)([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Magnesium (Mg)=<0.01 %
Nickel (Ni)=<0.001 %
Appearance (Form)Crystalline powder
Titration with NaOH>=98.5 %
pH4.1 to 4.7 (5 % solution)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only