Palladium(II) nitrate hydrate
Palladium(II) nitrate hydrate
Palladium(II) nitrate hydrate
Acros Organics

Palladium(II) nitrate hydrate

CAS: 207596-32-5 | N2O6Pd | 230.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number 389910250
HSN 28439019
Price (INR)
516,298.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
516,298.00
Each
Chemical Identifiers
CAS207596-32-5
IUPAC Namepalladium(2+) dinitrate
Molecular FormulaN2O6Pd
InChI KeyGPNDARIEYHPYAY-UHFFFAOYSA-N
SMILES[Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Appearance (Color)Brown to brown-black
Palladium (Pd)39 to 46 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only