IgA Mouse ELISA Standard
Product Image
Invitrogen™

IgA Mouse ELISA Standard

This product is a component of a respective eBioscience Kit. Please refer to the relevant manual for each product onRead more
Have Questions?
Catalog NumberQuantity
39-50450-654 pack
Catalog number 39-50450-65
Price (INR)
-
Quantity:
4 pack
This product is a component of a respective eBioscience Kit. Please refer to the relevant manual for each product on our website.

IgA comprises approximately 15% of all immunoglobulins in healthy human serum. IgA in serum is mainly monomeric, but in secretions, such as saliva, tears, colostrums, mucus, sweat, gastric fluid, IgA is found as a dimer where they are connected by a joining peptide. Most IgA is present in secreted form. This is believed to be due to ist properties in preventing invading pathogens by attaching and penetrating epithelial surfaces. IgA is just a very weak complement activating antibody; hence it does not induce bacterial cell lysis via the complement system. However secretory IgA works together with lysozymes, also present in many secreted fluids, which hydrolyze carbohydrates in bacterial cell walls enabling the immune system to clear the infection. IgA is predominantly found on epithelial cell surfaces where it acts as a neutralizing antibody.

Reported Application
ELISA, ELISA Recombinant Standard

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Microplate Reader
FormatKit
Former IdentifierseBioscience
Immunoassay Kit FormatELISA Standard
Quantity4 pack
Target Kit NamedIgA
Target Species NamedMouse
Target Species PrimaryMouse
Target Species ValidatedMouse
RecombinantRecombinant
Unit SizeEach